Assay Details
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139094.1 | NP_620794.1 | 16 - 17 | 2245 | 58 | |
XM_006227858.2 | XP_006227920.1 | 17 - 18 | 2172 | 58 | ||
GenBank mRNA | U49055.1 | - | 16 - 17 | 2245 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P13362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152926 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13362 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13362 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P13362 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13362 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700539971 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84930 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67922 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84115 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79372 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69983 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29133 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700539971 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10702761 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702762 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702756 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702758 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702759 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702753 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702767 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702748 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702765 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702749 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702747 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702752 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702754 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702757 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702764 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702755 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702763 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702750 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702751 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702766 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702760 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10702746 | - | - |
Rat Genome 230 2.0 Array | 1369896_PM_s_at | - | - |
Rat Genome 230 2.0 Array | 1377579_at | - | - |
Rat Genome 230 2.0 Array | 1377486_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377579_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369896_s_at | - | - |
Rat Genome 230 2.0 Array | 1377486_at | - | - |
Affymetrix HT Rat Focus Array | 1377486_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369896_s_at | - | - |
Affymetrix HT Rat Focus Array | 1377486_at | - | - |
Affymetrix HT Rat Focus Array | 1369896_PM_s_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100064670.rn.1 | TC0100008004.rn.1 | RTA TC |
Rat Expression Set 230 | 1377486_PM_at | - | - |
Rat Expression Set 230 | 1377486_at | - | - |
Rat Expression Set 230 | 1369896_s_at | - | - |
Rat Expression Set 230 | 1369896_PM_s_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702746 | - | - |
Rat Genome U34 Set | U49055_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22424556 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364407 | - | - |
RatRef-12 Expression BeadChip | ILMN_59332 | - | - |
