Assay Details
| Gene Symbol: Lrrc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145083.1 | NP_659551.1 | 1 | 47 | 71 | |
| XM_006248482.2 | XP_006248544.1 | 2 | 163 | 71 | ||
| GenBank mRNA | AB071036.1 | - | 1 | 47 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lrrc15
Genomic Map