Assay Details
| Gene Symbol: Npb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153293.1 | NP_695205.1 | 1 - 2 | 231 | 85 | |
| GenBank mRNA | AB085944.1 | - | 1 - 2 | 231 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npb
Genomic Map