Assay Details
| Gene Symbol: Dus1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172018.1 | NP_742015.1 | 11 - 12 | 1272 | 115 | |
| XM_006247874.1 | XP_006247936.1 | 11 - 12 | 1465 | 115 | ||
| XM_006247875.1 | XP_006247937.1 | 11 - 12 | 1405 | 115 | ||
| GenBank mRNA | AF508021.1 | - | 11 - 12 | 1272 | 115 | |
| AY327509.1 | - | 11 - 12 | 1272 | 115 | ||
| AY327510.1 | - | 11 - 12 | 1272 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dus1l
Genomic Map