Assay Details
| Gene Symbol: Elac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172326.1 | NP_758829.1 | 12 - 13 | 1131 | 100 | |
| GenBank mRNA | AY149902.1 | - | 12 - 13 | 1131 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P306316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92763 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P306316 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P306316 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P306316 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P306316 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P433367 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45760 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94891 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47061 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10734396 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734417 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734407 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734418 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734411 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734405 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734403 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734400 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734414 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734419 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734404 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734397 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734413 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734409 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734402 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734399 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734416 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734412 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734415 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734398 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734406 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734408 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734410 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734401 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10734395 | - | - |
| Rat Genome 230 2.0 Array | 1374107_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373581_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373581_at | - | - |
| Rat Genome 230 2.0 Array | 1374107_at | - | - |
| Affymetrix HT Rat Focus Array | 1374107_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374107_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000007007.rn.1 | TC1000000839.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000007008.rn.1 | TC1000000839.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374107_PM_at | - | - |
| Rat Expression Set 230 | 1374107_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734395 | - | - |
| Rat Genome U34 Set | rc_AI169160_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20965199 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361657 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62015 | - | - |

Elac2
Genomic Map