Assay Details
| Gene Symbol: Clic6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176078.2 | NP_788267.1 | 2 - 3 | 1566 | 89 | |
| GenBank mRNA | AY191249.1 | - | 2 - 3 | 1566 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clic6
Genomic Map