Assay Details
| Gene Symbol: Sh3bp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022693.4 | NP_073184.3 | 3 - 4 | 498 | 99 | |
| XM_008767298.1 | XP_008765520.1 | 1 - 2 | 1144 | 99 | ||
| GenBank mRNA | AJ278266.2 | - | 1 - 2 | 173 | 99 | |
| FM034920.1 | - | 3 - 4 | 498 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3bp4
Genomic Map