Assay Details
| Gene Symbol: Gdf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024375.1 | NP_077351.1 | 548 | 105 | ||
| GenBank mRNA | D49494.1 | - | 548 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gdf10
Genomic Map