Assay Details
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032617.2 | NP_116006.1 | 1 - 2 | 91 | 68 | |
GenBank mRNA | AF286534.1 | - | 1 - 2 | 97 | 68 | |
BC062041.1 | - | 1 - 2 | 91 | 5598168 | 68 | |
D01046.1 | - | 1 - 2 | 77 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
