Assay Details
| Gene Symbol: B3gat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054003.1 | NP_446455.1 | 3 - 4 | 339 | 67 | |
| XM_006242733.2 | XP_006242795.1 | 3 - 4 | 611 | 67 | ||
| XM_006242734.2 | XP_006242796.1 | 3 - 4 | 600 | 67 | ||
| GenBank mRNA | D88035.1 | - | 3 - 4 | 339 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

B3gat1
Genomic Map