Assay Details
| Gene Symbol: Cyp26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130408.2 | NP_569092.2 | 2 - 3 | 457 | 65 | |
| GenBank mRNA | AF439720.2 | - | 2 - 3 | 457 | 65 | |
| DQ317305.1 | - | 2 - 3 | 467 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp26a1
Genomic Map