Assay Details
Gene Symbol: Adrbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012897.2 | NP_037029.1 | 12 - 13 | 1059 | 73 | |
XM_008769317.1 | XP_008767539.1 | 11 - 12 | 950 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
