Assay Details
| Gene Symbol: Pam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013000.2 | NP_037132.2 | 7 - 8 | 769 | 61 | |
| XM_006245594.2 | XP_006245656.1 | 7 - 8 | 982 | 61 | ||
| XM_006245595.2 | XP_006245657.1 | 7 - 8 | 982 | 61 | ||
| XM_006245596.2 | XP_006245658.1 | 7 - 8 | 982 | 61 | ||
| XM_006245597.2 | XP_006245659.1 | 7 - 8 | 982 | 61 | ||
| XM_006245598.2 | XP_006245660.1 | 7 - 8 | 981 | 61 | ||
| XM_006245599.2 | XP_006245661.1 | 7 - 8 | 981 | 61 | ||
| XM_008767360.1 | XP_008765582.1 | 7 - 8 | 982 | 61 | ||
| GenBank mRNA | M25719.1 | - | 7 - 8 | 885 | 61 | |
| M25732.1 | - | 7 - 8 | 769 | 61 | ||
| M82845.1 | - | 7 - 8 | 640 | 61 | ||
| X59685.1 | - | 4 - 5 | 243 | 61 | ||
| X59686.1 | - | 4 - 5 | 239 | 61 | ||
| X59687.1 | - | 6 - 7 | 411 | 61 | ||
| X59688.1 | - | 6 - 7 | 400 | 61 | ||
| X59689.1 | - | 7 - 8 | 590 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pam
Genomic Map