Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139091.2 | NP_620791.1 | 4 - 5 | 615 | 75 | |
| XM_006252093.2 | XP_006252155.1 | 4 - 5 | 720 | 75 | ||
| GenBank mRNA | AF000900.1 | - | 4 - 5 | 552 | 75 | |
| AF000901.1 | - | 3 - 4 | 508 | 75 | ||
| BC085690.1 | - | 4 - 5 | 615 | 7190650 | 75 | |
| U63839.1 | - | 4 - 5 | 628 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF000901.1 transcripts.
Set Membership: |
|

Nupl1
Genomic Map