Assay Details
| Gene Symbol: Cd1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017079.1 | NP_058775.1 | 1 - 2 | 155 | 94 | |
| GenBank mRNA | D26439.2 | - | 1 - 2 | 155 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd1d1
Genomic Map