Assay Details
| Gene Symbol: Htr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017250.1 | NP_058946.1 | 1 - 2 | 578 | 81 | |
| XM_006245369.2 | XP_006245431.1 | 1 - 2 | 2281 | 81 | ||
| XM_008767216.1 | XP_008765438.1 | 2 - 3 | 1428 | 81 | ||
| GenBank mRNA | X66842.1 | - | 1 - 2 | 578 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Htr2b
Genomic Map