Assay Details
| Gene Symbol: Ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053881.1 | NP_446333.1 | 1 - 2 | 135 | 53 | |
| XM_006245134.2 | XP_006245196.1 | 1 - 2 | 203 | 53 | ||
| GenBank mRNA | FQ211652.1 | - | 1 - 2 | 194 | 53 | |
| U40652.1 | - | 1 - 2 | 101 | 53 | ||
| X92563.1 | - | 1 - 2 | 135 | 53 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptprn
Genomic Map