Assay Details
| Gene Symbol: Hs3st2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181370.2 | NP_852035.1 | 1 - 2 | 921 | 84 | |
| GenBank mRNA | AY240873.1 | - | 1 - 2 | 921 | 84 | |
| BC128721.1 | - | 1 - 2 | 924 | 8364509 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P780613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109746 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P780613 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P780613 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P780613 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P780613 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101473 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72121 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89776 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66090 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1846 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84495 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70953 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10710576 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710579 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710577 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10710575 | - | - |
| Rat Genome 230 2.0 Array | 1374046_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374046_at | - | - |
| Affymetrix HT Rat Focus Array | 1374046_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374046_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100019434.rn.1 | TC0100002518.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100019435.rn.1 | TC0100002518.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374046_at | - | - |
| Rat Expression Set 230 | 1374046_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710575 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21105273 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371347 | - | - |
| RatRef-12 Expression BeadChip | ILMN_48684 | - | - |

Hs3st2
Genomic Map