Assay Details
| Gene Symbol: Gldn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181382.2 | NP_852047.1 | 1 - 2 | 549 | 79 | |
| GenBank mRNA | AY266116.1 | - | 1 - 2 | 549 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P055137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77369 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055137 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055137 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P055137 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1011369 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43958 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93479 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27706 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53429 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10910055 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910050 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910057 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910056 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910054 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910053 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910052 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910051 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10910049 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800004843.rn.1 | TC0800000735.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800004844.rn.1 | TC0800000735.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800004846.rn.1 | TC0800000735.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10910049 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21132334 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373820 | - | - |
| RatRef-12 Expression BeadChip | ILMN_61557 | - | - |

Gldn
Genomic Map