Assay Details
| Gene Symbol: Kif15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181635.2 | NP_853666.1 | 6 - 7 | 488 | 145 | |
| GenBank mRNA | AY291580.1 | - | 6 - 7 | 488 | 145 | |
| AY291581.1 | - | 6 - 7 | 488 | 145 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P388755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51873 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P388755 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P388755 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P388755 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P388755 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92373 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94111 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51986 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40839 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52286 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1860 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10914493 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914504 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914492 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914494 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914502 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914490 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914485 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914509 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914516 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914512 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914515 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914483 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914486 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914499 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914491 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914496 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914488 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914501 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914489 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914484 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914500 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914498 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914505 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914503 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914497 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914487 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914513 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914495 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914514 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921092 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10914481 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10921090 | - | - |
| Rat Genome 230 2.0 Array | 1374794_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374794_at | - | - |
| Affymetrix HT Rat Focus Array | 1374794_at | - | - |
| Affymetrix HT Rat Focus Array | 1374794_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800014602.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014603.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014615.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014625.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014595.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014607.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014609.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014614.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014620.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014624.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014593.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014596.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014597.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014601.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014605.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014617.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014619.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014622.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014598.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014610.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014612.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014592.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014600.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014604.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014606.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014611.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014618.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014623.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014626.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014594.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014599.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014608.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014613.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014616.rn.1 | TC0800001825.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014621.rn.1 | TC0800001825.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374794_PM_at | - | - |
| Rat Expression Set 230 | 1374794_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914481 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22376368 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373916 | - | - |
| RatRef-12 Expression BeadChip | ILMN_51986 | - | - |

Kif15
Genomic Map