Assay Details
| Gene Symbol: Ccdc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182736.1 | NP_877400.1 | 10 - 11 | 895 | 129 | |
| XM_008768795.1 | XP_008767017.1 | 11 - 12 | 1660 | 129 | ||
| GenBank mRNA | AJ534984.1 | - | 10 - 11 | 895 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P540660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105885 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P540660 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P540660 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P540660 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P540660 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64875 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8450 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85088 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56583 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10755000 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754996 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754997 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755003 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755004 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754998 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755007 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755001 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10754995 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100011645.rn.1 | TC1100001832.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10754995 | - | - |
| Rat Genome U34 Set | rc_AI111898_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20760007 | - | - |
| ABI Rat Genome Survey Microarray | 21570896 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368724 | - | - |
| RatRef-12 Expression BeadChip | ILMN_64338 | - | - |

Ccdc50
Genomic Map