Assay Details
| Gene Symbol: Cyp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012941.2 | NP_037073.1 | 6 - 7 | 935 | 86 | |
| GenBank mRNA | AY724494.1 | - | 6 - 7 | 1257 | 86 | |
| BC087033.1 | - | 6 - 7 | 935 | 7130609 | 86 | |
| D55681.1 | - | 6 - 7 | 988 | 86 | ||
| U17697.1 | - | 6 - 7 | 1101 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cyp51
Genomic Map