Assay Details
| Gene Symbol: Ptbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271057.1 | NP_001257986.1 | 6 - 7 | 784 | 60 | |
| NM_022516.5 | NP_071961.2 | 6 - 7 | 784 | 60 | ||
| GenBank mRNA | BC061858.1 | - | 6 - 7 | 779 | 5598495 | 60 |
| CK365398.1 | - | 3 - 4 | 370 | 60 | ||
| X60789.1 | - | 6 - 7 | 747 | 60 | ||
| X60790.1 | - | 2 - 3 | 158 | 60 | ||
| X74565.1 | - | 6 - 7 | 765 | 60 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ptbp1
Genomic Map