Assay Details
Gene Symbol: Mif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031051.1 | NP_112313.1 | 1 - 2 | 192 | 101 | |
GenBank mRNA | BC061545.1 | - | 1 - 2 | 176 | 6920605 | 101 |
FQ210474.1 | - | 1 - 2 | 174 | 101 | ||
FQ217414.1 | - | 1 - 2 | 172 | 101 | ||
FQ218686.1 | - | 1 - 2 | 161 | 101 | ||
FQ222835.1 | - | 1 - 2 | 188 | 101 | ||
FQ223656.1 | - | 1 - 2 | 149 | 101 | ||
FQ229019.1 | - | 1 - 2 | 101 | |||
FQ229084.1 | - | 1 - 2 | 172 | 101 | ||
S73424.1 | - | 1 - 2 | 157 | 101 | ||
U62326.1 | - | 1 - 2 | 192 | 101 |
Gene Symbol: LOC103694877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773846.1 | XP_008772068.1 | 1 - 2 | 356 | 101 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
