Assay Details
| Gene Symbol: Rps24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031112.1 | NP_112374.1 | 2 - 3 | 97 | 67 | |
| XM_006252530.2 | XP_006252592.1 | 2 - 3 | 168 | 67 | ||
| XM_006252531.2 | XP_006252593.1 | 2 - 3 | 168 | 67 | ||
| XM_006252532.2 | XP_006252594.1 | 2 - 3 | 168 | 67 | ||
| XM_006252533.2 | XP_006252595.1 | 2 - 3 | 168 | 67 | ||
| GenBank mRNA | BC058140.1 | - | 2 - 3 | 108 | 6919886 | 67 |
| FQ221331.1 | - | 2 - 3 | 102 | 67 | ||
| FQ222238.1 | - | 2 - 3 | 102 | 67 | ||
| FQ223039.1 | - | 2 - 3 | 102 | 67 | ||
| FQ225772.1 | - | 2 - 3 | 102 | 67 | ||
| X51538.1 | - | 2 - 3 | 97 | 67 | ||
| X52445.1 | - | 2 - 3 | 97 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps24
Genomic Map