Assay Details
| Gene Symbol: Tdg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053729.1 | NP_446181.1 | 7 - 8 | 833 | 80 | |
| GenBank mRNA | AY026945.1 | - | 7 - 8 | 833 | 80 | |
| BC129088.1 | - | 7 - 8 | 850 | 8362408 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tdg
Genomic Map