Assay Details
| Gene Symbol: Cebpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301715.1 | NP_001288644.1 | 1 | 108 | 63 | |
| NM_001301720.1 | NP_001288649.1 | 1 | 108 | 63 | ||
| NM_024125.5 | NP_077039.3 | 1 | 108 | 63 | ||
| GenBank mRNA | BC129071.1 | - | 1 | 98 | 7458221 | 63 |
| M57235.1 | - | 1 | 95 | 63 | ||
| S77528.1 | - | 1 | 493 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001301715.1,NM_001301720.1 transcripts.
This assay is in the 5'UTR region of the NM_001301715.1,NM_001301720.1 transcripts.
Set Membership: |
|

Cebpb
Genomic Map