Assay Details
| Gene Symbol: Bcl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277288.1 | NP_001264217.1 | 2 - 3 | 426 | 66 | |
| XM_006240540.2 | XP_006240602.1 | 2 - 3 | 589 | 66 | ||
| GenBank mRNA | JF520428.1 | - | 2 - 3 | 426 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcl11b
Genomic Map