Assay Details
Gene Symbol: Atp5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134364.1 | NP_599191.1 | 4 - 5 | 628 | 83 | |
GenBank mRNA | BC099743.1 | - | 4 - 5 | 611 | 7937989 | 83 |
DQ403101.1 | - | 1 - 2 | 108 | 83 | ||
FQ214851.1 | - | 1 - 2 | 120 | 83 | ||
FQ216116.1 | - | 2 - 3 | 208 | 83 | ||
FQ230301.1 | - | 2 - 3 | 247 | 83 | ||
M19044.1 | - | 3 - 4 | 444 | 83 | ||
M57634.1 | - | 1 - 2 | 93 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
