Assay Details
| Gene Symbol: Osgin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_232798.8 | XP_232798.4 | 1 - 2 | 188 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_232798.8 transcripts.
Set Membership: |
|

Osgin2
Genomic Map