Assay Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033705.1 | NP_001028877.1 | 1 - 2 | 183 | 61 | |
| XM_008766953.1 | XP_008765175.1 | 1 - 2 | 209 | 61 | ||
| GenBank mRNA | BC101930.1 | - | 1 - 2 | 183 | 7938755 | 61 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC101930.1, NM_001033705.1, XM_008766953.1 transcripts.
Set Membership: |
|

Zfp451
Genomic Map