Assay Details
| Gene Symbol: Fam104a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034958.2 | NP_001030130.2 | 1 - 2 | 223 | 70 | |
| XM_006247695.2 | XP_006247757.1 | 1 - 2 | 243 | 70 | ||
| GenBank mRNA | BC087161.1 | - | 1 - 2 | 65 | 70 | |
| BC105876.1 | - | 1 - 2 | 94 | 7129874 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105876.1 transcripts.
Set Membership: |
|

Fam104a
Genomic Map