Assay Details
| Gene Symbol: Morf4l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007714.1 | NP_001007715.1 | 2 - 3 | 161 | 84 | |
| XM_006257295.1 | XP_006257357.1 | 2 - 3 | 197 | 84 | ||
| XM_006257296.2 | XP_006257358.1 | 2 - 3 | 191 | 84 | ||
| GenBank mRNA | BC083606.1 | - | 2 - 3 | 161 | 7133330 | 84 |
| FQ232538.1 | - | 2 - 3 | 160 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC083606.1,NM_001007714.1,XM_006257295.1,XM_006257296.2 transcripts.
This assay is in the 5'UTR region of the BC083606.1,NM_001007714.1,XM_006257295.1,XM_006257296.2 transcripts.
Set Membership: |
|

Morf4l2
Genomic Map