Assay Details
| Gene Symbol: Fam19a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191991.1 | NP_001178920.1 | 2 - 3 | 271 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam19a5| Gene Symbol: Fam19a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191991.1 | NP_001178920.1 | 2 - 3 | 271 | 95 | |
Set Membership: |
|