Assay Details
Gene Symbol: Upf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135873.1 | NP_001129345.1 | 4 - 5 | 577 | 113 | |
XM_006257455.2 | XP_006257517.1 | 4 - 5 | 600 | 113 | ||
XM_008773416.1 | XP_008771638.1 | 4 - 5 | 600 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
