Assay Details
| Gene Symbol: Stub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025625.2 | NP_001020796.2 | 2 - 3 | 460 | 58 | |
| GenBank mRNA | BC099240.1 | - | 2 - 3 | 407 | 7445727 | 58 |
| CB568781.1 | - | 2 - 3 | 461 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stub1
Genomic Map