Assay Details
| Gene Symbol: Ccdc120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191729.1 | NP_001178658.1 | 4 - 5 | 646 | 58 | |
| GenBank mRNA | FQ211363.1 | - | 889 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc120
Genomic Map