Assay Details
| Gene Symbol: Cdc42ep5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108469.1 | NP_001101939.1 | 1 | 467 | 60 | |
| XM_006228217.2 | XP_006228279.1 | 2 | 711 | 60 | ||
| XM_008758944.1 | XP_008757166.1 | 3 | 998 | 60 | ||
| XM_008758945.1 | XP_008757167.1 | 2 | 891 | 60 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cdc42ep5
Genomic Map