Assay Details
| Gene Symbol: Rqcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009357.1 | NP_001009357.1 | 1 - 2 | 442 | 111 | |
| GenBank mRNA | BC087134.1 | - | 1 - 2 | 442 | 7122272 | 111 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC087134.1, NM_001009357.1 transcripts.
Set Membership: |
|

Rqcd1
Genomic Map