Assay Details
| Gene Symbol: Rqcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009357.1 | NP_001009357.1 | 2 - 3 | 619 | 76 | |
| GenBank mRNA | BC087134.1 | - | 2 - 3 | 619 | 7122272 | 76 |
| FQ225078.1 | - | 2 - 3 | 334 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rqcd1
Genomic Map