Assay Details
Gene Symbol: Ctdsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128079.1 | NP_001121551.1 | 3 - 4 | 335 | 66 | |
XM_006245236.2 | XP_006245298.1 | 2 - 3 | 1108 | 66 | ||
GenBank mRNA | BC161976.1 | - | 3 - 4 | 335 | 9029838 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
