Assay Details
| Gene Symbol: Akp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022680.1 | NP_073171.1 | 6 - 7 | 781 | 64 | |
| GenBank mRNA | S66545.1 | - | 834 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akp3
Genomic Map