Assay Details
| Gene Symbol: Cyp27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245175.1 | XP_006245237.1 | 7 - 8 | 1152 | 91 | |
| GenBank mRNA | BC061848.1 | - | 7 - 8 | 1316 | 5621918 | 91 |
More Information
Set Membership: |
|

Cyp27a1
Genomic Map