Assay Details
| Gene Symbol: Map7d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271327.1 | NP_001258256.1 | 3 - 4 | 241 | 71 | |
| XM_008773649.1 | XP_008771871.1 | 3 - 4 | 437 | 71 | ||
| XM_008773650.1 | XP_008771872.1 | 3 - 4 | 349 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map7d3
Genomic Map