Assay Details
| Gene Symbol: Snapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013121.1 | NP_001013139.1 | 3 - 4 | 397 | 64 | |
| XM_008768956.1 | XP_008767178.1 | 3 - 4 | 500 | 64 | ||
| GenBank mRNA | BC079077.1 | - | 3 - 4 | 397 | 7122319 | 64 |
| FQ214278.1 | - | 2 - 3 | 227 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snapc2
Genomic Map