Assay Details
| Gene Symbol: Slc22a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108193.1 | NP_001101663.1 | 1374 | 63 | ||
| GenBank mRNA | BC168201.1 | - | 1409 | 7114743 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a14
Genomic Map