Assay Details
| Gene Symbol: Fam83b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108169.1 | NP_001101639.1 | 4 - 5 | 847 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P281661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170124 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P281661 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P281661 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P281661 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P281661 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32539 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10918771 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918773 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918774 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918772 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10918770 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800023710.rn.1 | TC0800002955.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10918770 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21282229 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367099 | - | - |

Fam83b
Genomic Map