Assay Details
| Gene Symbol: Med15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108325.1 | NP_001101795.1 | 13 - 14 | 1744 | 63 | |
| XM_006248702.2 | XP_006248764.1 | 12 - 13 | 1747 | 63 | ||
| XM_006248706.2 | XP_006248768.1 | 13 - 14 | 1867 | 63 | ||
| XM_008768891.1 | XP_008767113.1 | 12 - 13 | 1736 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Med15
Genomic Map