Assay Details
| Gene Symbol: Pcdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199503.1 | NP_955797.1 | 1 | 2337 | 68 | |
| GenBank mRNA | AB113384.1 | - | 1 | 2337 | 68 | |
| AY573971.1 | - | 1 | 2377 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P055669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73034 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055669 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055669 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P055669 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84886 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85341 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28130 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36527 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12051 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28208 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55436 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82784 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35049 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48373 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66078 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99914 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103120 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98248 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2823 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9647 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37198 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76629 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10890 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10801161 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10801139 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10801162 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10801159 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10801138 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10801160 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10801137 | - | - |
| Rat Genome 230 2.0 Array | 1390821_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393149_at | - | - |
| Rat Genome 230 2.0 Array | 1393149_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390821_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1800002556.rn.1 | TC1800000324.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390821_at | - | - |
| Rat Expression Set 230 | 1393149_PM_at | - | - |
| Rat Expression Set 230 | 1393149_at | - | - |
| Rat Expression Set 230 | 1390821_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10801137 | - | - |
| Rat Genome U34 Set | rc_AI170757_at | - | - |
| Rat Genome U34 Set | rc_AA819900_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21459852 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350900 | - | - |
| RatRef-12 Expression BeadChip | ILMN_69480 | - | - |

Pcdha1
Genomic Map