Assay Details
| Gene Symbol: Sim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107108.3 | NP_001100578.1 | 10 - 11 | 2135 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
signal transduction
embryonic pattern specification
lung development
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
protein heterodimerization activity
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
signal transduction
embryonic pattern specification
lung development
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P23237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2776 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P23237 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P23237 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P23237 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P23237 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P340513 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P457717 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 105007 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10750408 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750407 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750415 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750413 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750412 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750417 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750411 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750416 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750414 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750418 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750409 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750410 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10750406 | - | - |
| Rat Genome 230 2.0 Array | 1385722_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385722_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100001480.rn.1 | TC1100000318.rn.1 | RTA TC |
| Rat Expression Set 230 | 1385722_at | - | - |
| Rat Expression Set 230 | 1385722_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750406 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22062617 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361502 | - | - |

Sim2
Genomic Map